Anti-FGA Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: Innovative Research Inc
Certificates
About this item
Type: Primary
Antigen: FGA
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Pig
Specifications
- Catalog No:
- 103388-288
- 103384-734
- Antigen Symbol:
- FGA
- FGA
- Antigen Name:
- Fibrinogen
- Fibrinogen
- Antigen Synonyms:
- FGB, FGG
- FGB, FGG
- Antibody Type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Pig
- Pig
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 100514666, 100514354, 403164
- 100514666, 100514354, 403164
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Full length native protein (purified) (Porcine).
- Full length native protein (purified) (Porcine).
- Format:
- IgG fraction
- IgG fraction
- Form:
- Frozen liquid
- Frozen liquid
- Molecular Weight:
- 340 kDa
- 340 kDa
- Concentration:
- 10.0 mg/ml
- 10.0 mg/ml
- Storage Buffer:
- 0.05M Sodium Phosphate; 0.1M NaCl; 1mM EDTA; pH 6.6
- 0.05M Sodium Phosphate; 0.1M NaCl; 1mM EDTA; pH 6.6
- Storage Temperature:
- –70 °C; Protect from light
- –70 °C; Protect from light
- Shipping Temperature:
- –70 °C
- –70 °C