Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-CAT Rabbit Polyclonal Antibody
Anti-CAT Rabbit Polyclonal Antibody
Certificates
Anti-CAT Rabbit Polyclonal Antibody

Specifications

  • Catalog No:
  • 103386-246
  • 103384-640
  • Antigen Symbol:
  • CAT
  • CAT
  • Antigen Name:
  • Catalase
  • Catalase
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 847
  • 847
  • Isotype:
  • IgG
  • IgG
  • Western Blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Full length native protein (purified) (Human).
  • Full length native protein (purified) (Human).
  • Format:
  • IgG fraction
  • IgG fraction
  • Form:
  • Frozen liquid
  • Frozen liquid
  • Molecular Weight:
  • 230 kDa
  • 230 kDa
  • Concentration:
  • 10.0 mg/ml
  • 10.0 mg/ml
  • Storage Buffer:
  • PBS; 0.1% Azide
  • PBS; 0.1% Azide
  • Storage Temperature:
  • –70 °C
  • –70 °C
  • Shipping Temperature:
  • –70 °C
  • –70 °C
  • Tested Applications:
  • Immunoelectrophoresis
  • Immunoelectrophoresis

Specifications