Anti-CTSG Rabbit Polyclonal Antibody
Supplier: Innovative Research Inc
Certificates
About this item
Type: Primary
Antigen: CTSG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 103386-242
- 103384-636
- Antigen Symbol:
- CTSG
- CTSG
- Antigen Name:
- Cathepsin G
- Cathepsin G
- Antibody Type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 1511
- 1511
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Full length native protein (purified) (Human).
- Full length native protein (purified) (Human).
- Format:
- Ammonium sulfate fraction
- Ammonium sulfate fraction
- Form:
- liquid
- liquid
- Molecular Weight:
- 30 kDa
- 30 kDa
- Purification:
- Purified by fractionation
- Purified by fractionation
- Concentration:
- 10.0 mg/ml
- 10.0 mg/ml
- Storage Buffer:
- PBS; 0.1% Azide
- PBS; 0.1% Azide
- Storage Temperature:
- 4 °C
- 4 °C
- Shipping Temperature:
- 4 °C
- 4 °C
- Tested Applications:
- Immunoelectrophoresis
- Immunoelectrophoresis