Order Entry
United States
ContactUsLinkComponent
 
Phosphate Buffered Saline, 10X, Advanced Biomatrix
Catalog # 103701-212
CAS Number:  
Phosphate Buffered Saline, 10X, Advanced Biomatrix
Catalog # 103701-212
Supplier Number:  5076-100ML
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Phosphate buffered saline, 10X
  • Size:
    100 mL
  • Environmentally Preferable:
  • Cat. No.:
    103701-212

Specifications

About this item

Use With Type I Collagen to help form 3D hydrogels.

  • Sterility tested
  • Useful for 3D collagen hydrogel procedure
  • Sterile filtered
  • pH: Approx pH 7.0 to 7.6

Formulated to use in conjunction with other products such as Type I collagen for forming a gel.

PBS 10X is provided in a user-friendly packaging for use and storage. This product is sterile and supplied as a ready to use solution.

Caution: This product is for R&D use only and is not intended for human or other uses. Please consult the Material Safety Data Sheet for information regarding hazards and safe handling practices.