

Tributyl phosphate ≥99% (by GC), EMPROVE® EXPERT Ph. Eur., SAFC®
Supplier: MilliporeSigma
Certificates
About this item
CAS Number:126-73-8
:
Formula:(CH₃(CH₂)₃O)₃PO
MDL Number:MFCD00009436
:266.32 g/mol
Boiling Pt:289 °C Dec.
Melting Pt:–79 °C
Density:0.97 g/cm³
Flash Pt:146 °C
Storage Temperature:Ambient
EINECS:204-800-2
:GHS08
:GHS07
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Purity (GC):≥ 99 %
- Identity (IR-spectrum):passes test
- Appearance:passes test
- Acidity:passes test
- Chloride (Cl):≤ 200 ppm
- Heavy metals (as Pb):≤ 20 ppm
- Density (d 25/25):0.966 - 0.986
- Refractive index (n 25/D):1.4205 - 1.4225
- n-Butyl alcohol (GC):≤ 1000 ppm
- Other residual solvents (ICH Q3C):excluded by manufacturing process
- Related substances (Highest single impurity):≤ 0.1 %
- Related substances (Sum of all impurities):≤ 0.3 %
- Water (according to Karl Fischer):≤ 0.1 %
- Total aerobic microbial count (TAMC):≤ 1000 CFU/ml
- Total combined yeasts/moulds count (TYMC):≤ 100 CFU/ml
- Bacterial endotoxins:≤ 10 I.U./ml
Specification Test Results
Frequently Bought Together