Phenol Red Broth with Dulcitol, Hardy Diagnostics
Catalog # 76311-618
Supplier: Hardy Diagnostics
CAS Number:
Specifications
- Pack type:Tube
- Description:Phenol Red Broth with Dulcitol
- Size:10 ml
- Cat. No.:76311-618
- Tube size:16×125 mm
Specifications
About this item
For the differentiation of microorganisms by carbohydrate fermentation. Contains a durham tube for gas detection.