These Nanosphere™ size standards are excellent for use with any application that requires a NIST™ traceable size standard with a very narrow standard deviation.
- Ease of dispersion and colloidal stability
- Minimize response of analytical systems to shape effects
- Highly uniform polystyrene spheres calibrated within billionths of a meter (nanometers)
- Used in laser light scattering studies and colloidal systems research
- Reliable source for world class particle technology solutions
Ideal for the calibration of electron and atomic force microscopes. Conveniently check the sizes of bacteria, viruses, ribosomes and sub-cellular components.