Slide-Fix™ PPC Slide Jars with Caps, Caplugs
Catalog # MSPP-2225450G8
Supplier: CAPLUGS MS
Specifications
- Color:Natural
- Description:Slide jars with caps
- Material:ok
- Cat. No.:MSPP-2225450G8
- Packaging:10/bag; 120/carton
- Cap color:Green
Specifications
About this item
These sterile Slide-Fix™ slide jars are ideal for the preservation of pap smear slides, fine needle aspiration (FNA) biopsies, and other small samples.
- Sterilized by cobalt gamma radiation
- Specially manufactured for in-situ hybridization
- Outstanding performance in preserving and transporting pap smear slides
- Cap material:Polyethylene (PE)
The rigid polypropylene construction of slide jars provides fortress-like protection for up to four slides in transit.