PBS-TWEEN®, Phosphate Buffered Saline Tablets, MilliporeSigma
Product Details & Documents
Dissolving one tablet in 1 liter of deionized water yields 140 mM NaCl, 3 mM KCl, 0.05% TWEEN® 20 Detergent, 10 mM phosphate buffer, pH 7.4 at 25°C.
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Glass bottle
- Description:PBS-TWEEN® Tablets
- Size:10 Tablets