TempDex 720 Heat Protection Welding Glove, Mapa Professional
Catalog # 75795-070
Supplier: MAPA
Specifications
- Glove color:Orange,Black
- Glove size:11
- Ambidextrous/Hand-specific:Hand-Specific
- Disposable/Reusable:Reusable
- Material:Nitrile
- Length:9 - 11"
- Liner Material:Terry cloth
- Finish:Dots
- Cat. No.:75795-070
- Supplier no.:720121
Specifications
About this item
This glove is ideal for the handling of sharp and hot metals providing protection from both cuts and heat.
- Nitrile coating with dotted grip facilitates work in moderately oily environments
- Knitted aramide fibers ensures protection from cuts
- Comfortable terry-cloth insulation
- Abrasion resistance
- Durability
Certifications: CE Cat. II marked. Cut Resistance ANSI CUT A2. ISO 13997: 10,2N (970g). Thermal Risks EN 407: X2XXXX. Mechanical Hazards EN 388: 4343B.