ContactUsLinkComponent
KWIK-STIK 2 Pack Proteus mirabilis ATCC® 29906™*, Microbiologics
  75879-060
 :  
undefined
KWIK-STIK 2 Pack Proteus mirabilis ATCC® 29906™*, Microbiologics
  75879-060
 :  01215P
 :  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    Proteus mirabilis derived from ATCC® 29906™*
  • Cat. No.:
    75879-060
  • Supplier no.:
    01215P
Everything needed to grow reference cultures for QC testing is included in this one handy device.
  • Over 900 strains available
  • Ready-to-use format saves time and money
  • Refrigerated storage is easy and economical
  • Online Certificate of Analysis provides detailed strain information
  • Traceability to reference cultures ensures authenticity
  • Technical Support experts available for guidance
All-in-one design reduces the risk of contamination and strains are less than or equal to four passages from reference culture.
Available in a two pack, universally used for all types of microbiological control testing.
 : KWIK-STIK’s simple, all-inclusive design makes life easier for lab techs while reducing chances for errors. Each of it contains a qualitative lyophilized microorganism pellet, ampoule of hydrating fluid, and inoculating swab.