To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:His Capture Kit, type 2
- Cat. No.:103568-410
- Storage temperature:+2…+8 °C
- No. of reactions:16 immobilizations, 1600 regenerations
Specifications
About this item
Reagents for capture of histidine-tagged molecules as ligands in biomolecular interaction analysis. Designed for use with Biacore 8K+, Biacore 8K (larger kit component volumes than in His Capture Kit) but can be used with all other Biacore systems.
- Enables capture of histidine-tagged ligands
- Physiological conditions can be used during the coupling
- Optimized and verified assay minimizing assay development
It is designed for capture of histidine-tagged proteins to an anti-histidine antibody and provides an alternative to capture on nickel-chelated nitrilotriacetic (NTA) groups.
The anti-histidine antibody is a monoclonal antibody directed against polyhistidine tags in the C- or N-terminus of a protein. Depending on the nature of the interacting molecules and the micro-environment around the histidine-tag, the affinity of the capture antibodies will vary.