Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse Anti-body Capture Kit, Type-2, Cytiva
Mouse Anti-body Capture Kit, Type-2, Cytiva
Catalog # 103568-402
Supplier:  Cytiva
CAS Number:  
undefined
Mouse Anti-body Capture Kit, Type-2, Cytiva
Catalog # 103568-402
Supplier:  Cytiva
Supplier Number:  29215281
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Mouse Antibody Capture Kit, Type 2
  • Cat. No.:
    103568-402
  • No. of reactions:
    16 immobilizations, 1600 regenerations

Specifications

About this item

Kit with reagents for the capture of mouse antibodies, in component volumes larger than the standard mouse antibody capture kit.

  • Binds all IgG subclasses (IgG1, IgG2a, IgG2b, and IgG3), IgA and IgM
  • Very stable binding with no or very low dissociation of captured antibodies

Designed for use with Biacore, but can also be used with all other Biacore systems.

Mouse antibody capture kit, type 2 is intended for capture of mouse antibodies as ligands in various biomolecular interaction analyses. Anti-mouse antibodies are suitable for immobilization on carboxyl derivatized surfaces using amine coupling kit and the included immobilization buffer, while the regeneration solution is used for regeneration of the surface.

Anti-mouse antibodies consist of polyclonal rabbit anti-mouse immunoglobulin antibodies reacting with all IgG subclasses.