PureYield™ Plasmid Midiprep System, Promega
Catalog # PAA6740
Supplier: Promega Corporation
CAS Number:
Specifications
- Method / Format:Column (silica-based)
- Prep size:Midi
- Refine target molecule:Plasmid
- Sample type:Bacteria
- Target molecule:Plasmid
- Description:PureYield Plasmid Midiprep Start-Up Kit, 110V Electrical (100 preps, manifold, 4 elution devices and free vacuum pump)
- Size:1 each
- Storage Conditions:15 °C to 30 °C
- Description:PureYield Plasmid Midiprep Start-Up Kit
- Cat. No.:PAA6740
- Supplier no.:A6740
Specifications
About this item
Isolates high-quality plasmid DNA from bacterial cultures
- Up to 200 µg of plasmid from 50 mL culture
- Transfection-quality plasmid DNA in 30 minutes
- No tedious, gravity-drip columns or post-elution alcohol precipitation
Ordering information: Midiprep Systems include cell lysis solution, cell resuspension solution, column wash, endotoxin removal wash, neutralization solution, nuclease-free water, PureYield binding columns, PureYield clearing columns. Start-Up Kit also includes Welch™ Vacuum Pump, 110V; Vac-Man™ Laboratory Vacuum Manifold, 20-sample capacity and four Eluator™ Vacuum Elution Devices.