Black Latex Finger Cots, Calapro
: Calapro
Perfect for covering finger wounds or protecting fingers. Suggested use for Class I Category A static sensitive electronic devices above 1000 V.
- Thickness: 5 mil
- Overall length: 2.55 - 2.95"
- Static dissipative
- Rolled end