ContactUsLinkComponent
Micro-Touch® Micro-Thin™ Nitrile Examination Gloves
Micro-Touch® Micro-Thin™ Nitrile Examination Gloves
Supplier:  Ansell Healthcare
New Product
Certificates
undefined
Micro-Touch® Micro-Thin™ Nitrile Examination Gloves
Supplier:  Ansell Healthcare
New Product
Specifications
  • Catalog No:
  • 75839-364
  • 75839-360
  • 75839-358
  • 77661-322
  • 75839-362
  • Ambidextrous/Hand-specific:
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Disposable/Reusable:
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Latex/Latex-free:
  • Latex-Free
  • Latex-Free
  • Latex-Free
  • Latex-Free
  • Latex-Free
  • Powdered/Powder-free:
  • Powder-Free
  • Powder-Free
  • Powder-Free
  • Powder-Free
  • Powder-Free
  • Finish:
  • Textured fingers
  • Textured fingers
  • Textured fingers
  • Textured fingers
  • Textured fingers
  • Cuff Style:
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Cuff Length:
  • Standard
  • Standard
  • Standard
  • Standard
  • Standard
  • Finger Thickness:
  • 3.9 mil
  • 3.9 mil
  • 3.9 mil
  • 3.9 mil
  • 3.9 mil
  • Palm Thickness:
  • 2.4 mil
  • 2.4 mil
  • 2.4 mil
  • 2.4 mil
  • 2.4 mil
  • FDA Regulated:
  • +
  • +
  • +
  • +
  • +
  • Sterility:
  • Non sterile
  • Non sterile
  • Non sterile
  • Non sterile
  • Non sterile
  • Food Contact Approved:
  • +
  • +
  • +
  • +
  • +
  • ASTM No.:
  • D6319
  • D6319
  • D6319
  • D6319
  • D6319
Specifications