S.F.P. Supplement (Perfringens S.F.P. Supplement), HiMedia
Supplier: HiMedia
Certificates
About this item
An antibiotic supplement, recommended for the selective isolation of C. perfringens.
- Store at 2 to 8 °C
Caution: Not for medicinal Use.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.