ContactUsLinkComponent
Ugo Basile Dynamic Plantar, Stoelting
Ugo Basile Dynamic Plantar, Stoelting
  100231-404
 :  Stoelting
Minority or Woman-Owned Business Enterprise
Ugo Basile Dynamic Plantar, Stoelting
  100231-404
 :  Stoelting
 :  57820
Minority or Woman-Owned Business Enterprise
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    Ugo basile dynamic plantar (Von Frey) for rodents
  • Cat. No.:
    100231-404
  • Supplier no.:
    57820
The ugo basile dynamic plantar aesthesiometer has been designed to automate the assessment of touch sensitivity on the plantar surface of rats or mice and comes ready to work with both mice and rats. Similar to electronic Von Frey or Plantar Von Frey instrument, but it differs from Von Frey devices as latency and actual force at the time of paw withdrawal reflex are automatically detected and recorded.
  • Movable touch-stimulator unit well proven in revious model
  • Large testing surface
  • Multifunction liquid-crystal graphic-display
  • New modular animal enclosure, offering from 3 to 12 spaces
The unit comes with a microprocessor controlled electronic unit, of new design provided with graphic display, internal memory for data storage, memory stick and optional printer.
DELTA 9-pin connector
85 - 264 V, 50 - 60 Hz
10.2 kg