ContactUsLinkComponent
Carrousel Pipette Stand, Greiner Bio-One
Catalog # 76299-826
Supplier:  Greiner Bio-One
undefined
Carrousel Pipette Stand, Greiner Bio-One
Catalog # 76299-826
Supplier:  Greiner Bio-One
Supplier Number:  89000099
Specifications
  • Description:
    Carrousel pipette stand
  • Cat. No.:
    76299-826
Specifications
About this item
This stand holds up to seven single and multi-channel pipettes.
  • Heavy base for stability
  • Robust and space-saving
  • Holds single and multi-channel pipettes