Orders LinkContactUsLinkComponent
SRT® SEC HPLC Column, Sepax Technologies
SRT® SEC HPLC Column, Sepax Technologies
Marketsource
Supplier:   Sepax Technologies
SRT® SEC HPLC Column, Sepax Technologies
Supplier:  Sepax Technologies
Marketsource
Certificates
SRT® SEC HPLC Column, Sepax Technologies
60 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
SRT® SEC phase, developed based on innovative surface coating technology, comprises of uniform, hydrophilic, and neutral nanometer thick film chemically bonded to high purity and mechanically stabilized silica.
  • High separation efficiency and resolution
  • High capacity
  • High lot-to-lot reproducibility
  • High protein recovery with intact biological activity
  • Negligible non-specific interactions
SRT® uses stand-up monolayer on porous silica as oppose to SRT®-C of lay-down monolayer, offers ideal phase for monoclonal antibodies, proteins, peptides, nucleic acids, oligonucleotides, virus and water-soluble polymers.
Product Family Options
Product Information
  • Column Format
    Phase
    Pore size
    Particle Size
    Ø int.
    Length
    Packing material
    Availability
    Price
  • Analytical
    SRT® SEC
    100 Å
    5 µm
    4.60 mm
    150 mm
    Chemically bonded and hydrophilic silica
    Log in for availability
    Log in for availability
    Each
    Each - $2,107.69
    $2,107.69
    Each
       $2,107.69
    Supplier Number: 215100-4615
    Specifications:
    Marketsource
    Marketsource
  • Analytical
    SRT® SEC
    100 Å
    5 µm
    4.60 mm
    250 mm
    Chemically bonded and hydrophilic silica
    Log in for availability
    Log in for availability
    Each
    Each - $2,480.00
    $2,480.00
    Each
       $2,480.00
    Supplier Number: 215100-4625
    Specifications:
    Marketsource
    Marketsource
  • Analytical
    SRT® SEC
    100 Å
    5 µm
    4.60 mm
    300 mm
    Chemically bonded and hydrophilic silica
    Log in for availability
    Log in for availability
    Each
    Each - $2,615.38
    $2,615.38
    Each
       $2,615.38
    Supplier Number: 215100-4630
    Specifications:
    Marketsource
    Marketsource
  • Analytical
    SRT® SEC
    100 Å
    5 µm
    4.60 mm
    300 mm
    PEEK
    Log in for availability
    Log in for availability
    Each
    Each - $2,887.69
    $2,887.69
    Each
       $2,887.69
    Supplier Number: 215100P-4630
    Specifications:
    Marketsource
    Marketsource
  • Analytical
    SRT® SEC
    150 Å
    5 µm
    4.60 mm
    150 mm
    Chemically bonded and hydrophilic silica
    Log in for availability
    Log in for availability
    Each
    Each - $2,107.69
    $2,107.69
    Each
       $2,107.69
    Supplier Number: 215150-4615
    Specifications:
    Marketsource
    Marketsource
  • Analytical
    SRT® SEC
    150 Å
    5 µm
    4.60 mm
    250 mm
    Chemically bonded and hydrophilic silica
    Log in for availability
    Log in for availability
    Each
    Each - $2,480.00
    $2,480.00
    Each
       $2,480.00
    Supplier Number: 215150-4625
    Specifications:
    Marketsource
    Marketsource
  • Analytical
    SRT® SEC
    150 Å
    5 µm
    4.60 mm
    300 mm
    Chemically bonded and hydrophilic silica
    Log in for availability
    Log in for availability
    Each
    Each - $2,615.38
    $2,615.38
    Each
       $2,615.38
    Supplier Number: 215150-4630
    Specifications:
    Marketsource
    Marketsource
  • Analytical
    SRT® SEC
    150 Å
    5 µm
    4.60 mm
    300 mm
    PEEK
    Log in for availability
    Log in for availability
    Each
    Each - $2,887.69
    $2,887.69
    Each
       $2,887.69
    Supplier Number: 215150P-4630
    Specifications:
    Marketsource
    Marketsource
  • Analytical
    SRT® SEC
    300 Å
    5 µm
    4.60 mm
    150 mm
    Chemically bonded and hydrophilic silica
    Log in for availability
    Log in for availability
    Each
    Each - $2,107.69
    $2,107.69
    Each
       $2,107.69
    Supplier Number: 215300-4615
    Specifications:
    Marketsource
    Marketsource
  • Analytical
    SRT® SEC
    300 Å
    5 µm
    4.60 mm
    250 mm
    Chemically bonded and hydrophilic silica
    Log in for availability
    Log in for availability
    Each
    Each - $2,480.00
    $2,480.00
    Each
       $2,480.00
    Supplier Number: 215300-4625
    Specifications:
    Marketsource
    Marketsource
  • Analytical
    SRT® SEC
    300 Å
    5 µm
    4.60 mm
    300 mm
    Chemically bonded and hydrophilic silica
    Log in for availability
    Log in for availability
    Each
    Each - $2,615.38
    $2,615.38
    Each
       $2,615.38
    Supplier Number: 215300-4630
    Specifications:
    Marketsource
    Marketsource
  • Analytical
    SRT® SEC
    300 Å
    5 µm
    4.60 mm
    300 mm
    PEEK
    Log in for availability
    Log in for availability
    Each
    Each - $2,887.69
    $2,887.69
    Each
       $2,887.69
    Supplier Number: 215300P-4630
    Specifications:
    Marketsource
    Marketsource
  • Analytical
    SRT® SEC
    500 Å
    5 µm
    4.60 mm
    150 mm
    Chemically bonded and hydrophilic silica
    Log in for availability
    Log in for availability
    Each
    Each - $2,146.15
    $2,146.15
    Each
       $2,146.15
    Supplier Number: 215500-4615
    Specifications:
    Marketsource
    Marketsource
  • Analytical
    SRT® SEC
    500 Å
    5 µm
    4.60 mm
    250 mm
    Chemically bonded and hydrophilic silica
    Log in for availability
    Log in for availability
    Each
    Each - $2,536.92
    $2,536.92
    Each
       $2,536.92
    Supplier Number: 215500-4625
    Specifications:
    Marketsource
    Marketsource
  • Analytical
    SRT® SEC
    500 Å
    5 µm
    4.60 mm
    300 mm
    Chemically bonded and hydrophilic silica
    Log in for availability
    Log in for availability
    Each
    Each - $2,770.77
    $2,770.77
    Each
       $2,770.77
    Supplier Number: 215500-4630
    Specifications:
    Marketsource
    Marketsource
  • Analytical
    SRT® SEC
    500 Å
    5 µm
    4.60 mm
    300 mm
    PEEK
    Log in for availability
    Log in for availability
    Each
    Each - $3,043.08
    $3,043.08
    Each
       $3,043.08
    Supplier Number: 215500P-4630
    Specifications:
    Marketsource
    Marketsource
  • Analytical
    SRT® SEC
    1000 Å
    5 µm
    4.60 mm
    150 mm
    Chemically bonded and hydrophilic silica
    Log in for availability
    Log in for availability
    Each
    Each - $2,146.15
    $2,146.15
    Each
       $2,146.15
    Supplier Number: 215950-4615
    Specifications:
    Marketsource
    Marketsource
  • Analytical
    SRT® SEC
    1000 Å
    5 µm
    4.60 mm
    250 mm
    Chemically bonded and hydrophilic silica
    Log in for availability
    Log in for availability
    Each
    Each - $2,536.92
    $2,536.92
    Each
       $2,536.92
    Supplier Number: 215950-4625
    Specifications:
    Marketsource
    Marketsource
  • Analytical
    SRT® SEC
    1000 Å
    5 µm
    4.60 mm
    300 mm
    Chemically bonded and hydrophilic silica
    Log in for availability
    Log in for availability
    Each
    Each - $2,770.77
    $2,770.77
    Each
       $2,770.77
    Supplier Number: 215950-4630
    Specifications:
    Marketsource
    Marketsource
  • Analytical
    SRT® SEC
    1000 Å
    5 µm
    4.60 mm
    300 mm
    PEEK
    Log in for availability
    Log in for availability
    Each
    Each - $3,043.08
    $3,043.08
    Each
       $3,043.08
    Supplier Number: 215950P-4630
    Specifications:
    Marketsource
    Marketsource
  • Analytical
    SRT® SEC
    2000 Å
    5 µm
    4.60 mm
    150 mm
    Chemically bonded and hydrophilic silica
    Log in for availability
    Log in for availability
    Each
    Each - $2,146.15
    $2,146.15
    Each
       $2,146.15
    Supplier Number: 215980-4615
    Specifications:
    Marketsource
    Marketsource
  • Analytical
    SRT® SEC
    2000 Å
    5 µm
    4.60 mm
    250 mm
    Chemically bonded and hydrophilic silica
    Log in for availability
    Log in for availability
    Each
    Each - $2,536.92
    $2,536.92
    Each
       $2,536.92
    Supplier Number: 215980-4625
    Specifications:
    Marketsource
    Marketsource
  • Analytical
    SRT® SEC
    2000 Å
    5 µm
    4.60 mm
    300 mm
    Chemically bonded and hydrophilic silica
    Log in for availability
    Log in for availability
    Each
    Each - $2,770.77
    $2,770.77
    Each
       $2,770.77
    Supplier Number: 215980-4630
    Specifications:
    Marketsource
    Marketsource
  • Analytical
    SRT® SEC
    2000 Å
    5 µm
    4.60 mm
    300 mm
    PEEK
    Log in for availability
    Log in for availability
    Each
    Each - $3,043.08
    $3,043.08
    Each
       $3,043.08
    Supplier Number: 215980P-4630
    Specifications:
    Marketsource
    Marketsource
  • Analytical
    SRT® SEC
    100 Å
    5 µm
    7.80 mm
    150 mm
    Chemically bonded and hydrophilic silica
    Log in for availability
    Log in for availability
    Each
    Each - $2,323.08
    $2,323.08
    Each
       $2,323.08
    Supplier Number: 215100-7815
    Specifications:
    Marketsource
    Marketsource
  • Analytical
    SRT® SEC
    100 Å
    5 µm
    7.80 mm
    250 mm
    Chemically bonded and hydrophilic silica
    Log in for availability
    Log in for availability
    Each
    Each - $2,615.38
    $2,615.38
    Each
       $2,615.38
    Supplier Number: 215100-7825
    Specifications:
    Marketsource
    Marketsource
  • Analytical
    SRT® SEC
    100 Å
    5 µm
    7.80 mm
    300 mm
    Chemically bonded and hydrophilic silica
    Log in for availability
    Log in for availability
    Each
    Each - $3,180.00
    $3,180.00
    Each
       $3,180.00
    Supplier Number: 215100-7830
    Specifications:
    Marketsource
    Marketsource
  • Analytical
    SRT® SEC
    150 Å
    5 µm
    7.80 mm
    150 mm
    Chemically bonded and hydrophilic silica
    Log in for availability
    Log in for availability
    Each
    Each - $2,323.08
    $2,323.08
    Each
       $2,323.08
    Supplier Number: 215150-7815
    Specifications:
    Marketsource
    Marketsource
  • Analytical
    SRT® SEC
    150 Å
    5 µm
    7.80 mm
    250 mm
    Chemically bonded and hydrophilic silica
    Log in for availability
    Log in for availability
    Each
    Each - $2,615.38
    $2,615.38
    Each
       $2,615.38
    Supplier Number: 215150-7825
    Specifications:
    Marketsource
    Marketsource
  • Analytical
    SRT® SEC
    150 Å
    5 µm
    7.80 mm
    300 mm
    Chemically bonded and hydrophilic silica
    Log in for availability
    Log in for availability
    Each
    Each - $3,180.00
    $3,180.00
    Each
       $3,180.00
    Supplier Number: 215150-7830
    Specifications:
    Marketsource
    Marketsource
  • Analytical
    SRT® SEC
    300 Å
    5 µm
    7.80 mm
    150 mm
    Chemically bonded and hydrophilic silica
    Log in for availability
    Log in for availability
    Each
    Each - $2,323.08
    $2,323.08
    Each
       $2,323.08
    Supplier Number: 215300-7815
    Specifications:
    Marketsource
    Marketsource
  • Analytical
    SRT® SEC
    300 Å
    5 µm
    7.80 mm
    250 mm
    Chemically bonded and hydrophilic silica
    Log in for availability
    Log in for availability
    Each
    Each - $2,615.38
    $2,615.38
    Each
       $2,615.38
    Supplier Number: 215300-7825
    Specifications:
    Marketsource
    Marketsource
  • Analytical
    SRT® SEC
    300 Å
    5 µm
    7.80 mm
    300 mm
    Chemically bonded and hydrophilic silica
    Log in for availability
    Log in for availability
    Each
    Each - $3,180.00
    $3,180.00
    Each
       $3,180.00
    Supplier Number: 215300-7830
    Specifications:
    Marketsource
    Marketsource
  • Analytical
    SRT® SEC
    500 Å
    5 µm
    7.80 mm
    150 mm
    Chemically bonded and hydrophilic silica
    Log in for availability
    Log in for availability
    Each
    Each - $2,595.38
    $2,595.38
    Each
       $2,595.38
    Supplier Number: 215500-7815
    Specifications:
    Marketsource
    Marketsource
  • Analytical
    SRT® SEC
    500 Å
    5 µm
    7.80 mm
    250 mm
    Chemically bonded and hydrophilic silica
    Log in for availability
    Log in for availability
    Each
    Each - $3,103.08
    $3,103.08
    Each
       $3,103.08
    Supplier Number: 215500-7825
    Specifications:
    Marketsource
    Marketsource
  • Analytical
    SRT® SEC
    500 Å
    5 µm
    7.80 mm
    300 mm
    Chemically bonded and hydrophilic silica
    Log in for availability
    Log in for availability
    Each
    Each - $3,395.38
    $3,395.38
    Each
       $3,395.38
    Supplier Number: 215500-7830
    Specifications:
    Marketsource
    Marketsource
  • Analytical
    SRT® SEC
    1000 Å
    5 µm
    7.80 mm
    150 mm
    Chemically bonded and hydrophilic silica
    Log in for availability
    Log in for availability
    Each
    Each - $2,595.38
    $2,595.38
    Each
       $2,595.38
    Supplier Number: 215950-7815
    Specifications:
    Marketsource
    Marketsource
  • Analytical
    SRT® SEC
    1000 Å
    5 µm
    7.80 mm
    250 mm
    Chemically bonded and hydrophilic silica
    Log in for availability
    Log in for availability
    Each
    Each - $3,103.08
    $3,103.08
    Each
       $3,103.08
    Supplier Number: 215950-7825
    Specifications:
    Marketsource
    Marketsource
  • Analytical
    SRT® SEC
    1000 Å
    5 µm
    7.80 mm
    300 mm
    Chemically bonded and hydrophilic silica
    Log in for availability
    Log in for availability
    Each
    Each - $3,395.38
    $3,395.38
    Each
       $3,395.38
    Supplier Number: 215950-7830
    Specifications:
    Marketsource
    Marketsource
  • Analytical
    SRT® SEC
    2000 Å
    5 µm
    7.80 mm
    150 mm
    Chemically bonded and hydrophilic silica
    Log in for availability
    Log in for availability
    Each
    Each - $2,595.38
    $2,595.38
    Each
       $2,595.38
    Supplier Number: 215980-7815
    Specifications:
    Marketsource
    Marketsource
  • Analytical
    SRT® SEC
    2000 Å
    5 µm
    7.80 mm
    250 mm
    Chemically bonded and hydrophilic silica
    Log in for availability
    Log in for availability
    Each
    Each - $3,103.08
    $3,103.08
    Each
       $3,103.08
    Supplier Number: 215980-7825
    Specifications:
    Marketsource
    Marketsource
  • Analytical
    SRT® SEC
    2000 Å
    5 µm
    7.80 mm
    300 mm
    Chemically bonded and hydrophilic silica
    Log in for availability
    Log in for availability
    Each
    Each - $3,395.38
    $3,395.38
    Each
       $3,395.38
    Supplier Number: 215980-7830
    Specifications:
    Marketsource
    Marketsource
Guard Columns
Product Information
  • Column Format
    Phase
    Pore size
    Particle Size
    Ø int.
    Length
    Packing material
    Availability
    Price
  • Guard column
    SRT® SEC
    100 Å
    5 µm
    4.60 mm
    50 mm
    PEEK
    Log in for availability
    Log in for availability
    Each
    Each - $1,330.77
    $1,330.77
    Each
       $1,330.77
    Supplier Number: 215100P-4605
    Specifications:
    Marketsource
    Marketsource
  • Guard column
    SRT® SEC
    100 Å
    5 µm
    4.60 mm
    50 mm
    Chemically bonded and hydrophilic silica
    Log in for availability
    Log in for availability
    Each
    Each - $1,138.46
    $1,138.46
    Each
       $1,138.46
    Supplier Number: 215100-4605
    Specifications:
    Marketsource
    Marketsource
  • Guard column
    SRT® SEC
    150 Å
    5 µm
    4.60 mm
    50 mm
    PEEK
    Log in for availability
    Log in for availability
    Each
    Each - $1,330.77
    $1,330.77
    Each
       $1,330.77
    Supplier Number: 215150P-4605
    Specifications:
    Marketsource
    Marketsource
  • Guard column
    SRT® SEC
    150 Å
    5 µm
    4.60 mm
    50 mm
    Chemically bonded and hydrophilic silica
    Log in for availability
    Log in for availability
    Each
    Each - $1,138.46
    $1,138.46
    Each
       $1,138.46
    Minimum order quantity is 1
    Supplier Number: 215150-4605
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    Marketsource
    Marketsource
  • Guard column
    SRT® SEC
    300 Å
    5 µm
    4.60 mm
    50 mm
    PEEK
    Log in for availability
    Log in for availability
    Each
    Each - $1,330.77
    $1,330.77
    Each
       $1,330.77
    Supplier Number: 215300P-4605
    Specifications:
    Marketsource
    Marketsource
  • Guard column
    SRT® SEC
    300 Å
    5 µm
    4.60 mm
    50 mm
    Chemically bonded and hydrophilic silica
    Log in for availability
    Log in for availability
    Each
    Each - $1,138.46
    $1,138.46
    Each
       $1,138.46
    Supplier Number: 215300-4605
    Specifications:
    Marketsource
    Marketsource
  • Guard column
    SRT® SEC
    500 Å
    5 µm
    4.60 mm
    50 mm
    PEEK
    Log in for availability
    Log in for availability
    Each
    Each - $1,373.85
    $1,373.85
    Each
       $1,373.85
    Supplier Number: 215500P-4605
    Specifications:
    Marketsource
    Marketsource
  • Guard column
    SRT® SEC
    500 Å
    5 µm
    4.60 mm
    50 mm
    Chemically bonded and hydrophilic silica
    Log in for availability
    Log in for availability
    Each
    Each - $1,161.54
    $1,161.54
    Each
       $1,161.54
    Supplier Number: 215500-4605
    Specifications:
    Marketsource
    Marketsource
  • Guard column
    SRT® SEC
    1000 Å
    5 µm
    4.60 mm
    50 mm
    PEEK
    Log in for availability
    Log in for availability
    Each
    Each - $1,373.85
    $1,373.85
    Each
       $1,373.85
    Supplier Number: 215950P-4605
    Specifications:
    Marketsource
    Marketsource
  • Guard column
    SRT® SEC
    1000 Å
    5 µm
    4.60 mm
    50 mm
    Chemically bonded and hydrophilic silica
    Log in for availability
    Log in for availability
    Each
    Each - $1,161.54
    $1,161.54
    Each
       $1,161.54
    Supplier Number: 215950-4605
    Specifications:
    Marketsource
    Marketsource
  • Guard column
    SRT® SEC
    2000 Å
    5 µm
    4.60 mm
    50 mm
    PEEK
    Log in for availability
    Log in for availability
    Each
    Each - $1,373.85
    $1,373.85
    Each
       $1,373.85
    Supplier Number: 215980P-4605
    Specifications:
    Marketsource
    Marketsource
  • Guard column
    SRT® SEC
    2000 Å
    5 µm
    4.60 mm
    50 mm
    Chemically bonded and hydrophilic silica
    Log in for availability
    Log in for availability
    Each
    Each - $1,161.54
    $1,161.54
    Each
       $1,161.54
    Supplier Number: 215980-4605
    Specifications:
    Marketsource
    Marketsource
  • Guard column
    SRT® SEC
    100 Å
    5 µm
    7.80 mm
    50 mm
    Chemically bonded and hydrophilic silica
    Log in for availability
    Log in for availability
    Each
    Each - $1,138.46
    $1,138.46
    Each
       $1,138.46
    Supplier Number: 215100-7805
    Specifications:
    Marketsource
    Marketsource
  • Guard column
    SRT® SEC
    150 Å
    5 µm
    7.80 mm
    50 mm
    Chemically bonded and hydrophilic silica
    Log in for availability
    Log in for availability
    Each
    Each - $1,138.46
    $1,138.46
    Each
       $1,138.46
    Minimum order quantity is 1
    Supplier Number: 215150-7805
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    Marketsource
    Marketsource
  • Guard column
    SRT® SEC
    300 Å
    5 µm
    7.80 mm
    50 mm
    Chemically bonded and hydrophilic silica
    Log in for availability
    Log in for availability
    Each
    Each - $1,138.46
    $1,138.46
    Each
       $1,138.46
    Supplier Number: 215300-7805
    Specifications:
    Marketsource
    Marketsource
  • Guard column
    SRT® SEC
    500 Å
    5 µm
    7.80 mm
    50 mm
    Chemically bonded and hydrophilic silica
    Log in for availability
    Log in for availability
    Each
    Each - $1,203.08
    $1,203.08
    Each
       $1,203.08
    Supplier Number: 215500-7805
    Specifications:
    Marketsource
    Marketsource
  • Guard column
    SRT® SEC
    1000 Å
    5 µm
    7.80 mm
    50 mm
    Chemically bonded and hydrophilic silica
    Log in for availability
    Log in for availability
    Each
    Each - $1,203.08
    $1,203.08
    Each
       $1,203.08
    Supplier Number: 215950-7805
    Specifications:
    Marketsource
    Marketsource
  • Guard column
    SRT® SEC
    2000 Å
    5 µm
    7.80 mm
    50 mm
    Chemically bonded and hydrophilic silica
    Log in for availability
    Log in for availability
    Each
    Each - $1,203.08
    $1,203.08
    Each
       $1,203.08
    Supplier Number: 215980-7805
    Specifications:
    Marketsource
    Marketsource