Order Entry
Puerto Rico
ContactUsLinkComponent
Nrf2 Transcription Factor Assay Kit, Cayman
Nrf2 Transcription Factor Assay Kit, Cayman
Catalog # 75817-034
CAS Number:  
undefined
Nrf2 Transcription Factor Assay Kit, Cayman
Catalog # 75817-034
Supplier Number:  600590-96
CAS Number:  

Specifications

  • Storage Temperature:
    −80 °C
  • Cat. No.:
    75817-034
  • Supplier no.:
    600590-96
  • No. of tests:
    96 wells

Specifications

About this item

A 96-well assay for measurement of Nrf2.

  • A sensitive, non-radioactive method of detecting Nrf2 from whole cell lysates
  • 96-well plate format replaces EMSAs
  • Capture the transcription factor using a specific dsDNA sequence bound to the plate
  • Detect the dsDNA-bound transcription factor with specific antibodies in an ELISA format

Nuclear factor E2-related factor 2 (Nrf2) is a transcription factor that plays a key role in maintaining redox homeostasis via its interaction with a cysteine-rich protein Kelch-like ECH-associated protein 1 (Keap1). In resting cells, Nrf2 and Keap1 form a tight complex, which is targeted for degradation by proteasomes.

Under oxidative stress, Nrf2 is released from the Nrf2/Keap1 complex and translocate to the nucleus where it is able to induce the expression of a battery of genes encoding diverse cytoprotective proteins, including antioxidative enzymes, anti-inflammatory mediators, and proteasomes. Nrf2 Transcription Factor Assay is a non-radioactive, colorimetric method for detecting specific transcription factor DNA binding activity in nuclear extracts.

A specific double stranded DNA (dsDNA) sequence containing the Nrf2 response element is immobilized onto the wells of a 96-well plate. Nrf2 contained in nuclear extract samples will bind specifically to the Nrf2 response element and can be detected by addition of a specific Nrf2 antibody and a secondary antibody conjugated to HRP.

Delivery: Kit contains assay buffer, Reagent A, positive control, binding buffer, antibody, concentrate, polysorbate 20, competitor dsDNA, grp conjugate, plate and cover, developing and stop solution.