Order Entry
Puerto Rico
ContactUsLinkComponent
Scotch-Weld™ Epoxy Adhesive 1469, 3M™
Scotch-Weld™ Epoxy Adhesive 1469, 3M™
  MSPP-1469
 :  
Scotch-Weld™ Epoxy Adhesive 1469, 3M™
  MSPP-1469
 :  1469
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Color:
    Cream
  • Description:
    Scotch-Weld™ Epoxy Adhesive 1469
  • Cat. No.:
    MSPP-1469
  • Supplier no.:
    1469

 

 

This epoxy adhesive is a one-part, thermosetting liquid adhesive that provides exceptionally high strength at temperatures from –70 to 300 °F (–57 to 149 °C). This white to cream colored adhesive can be applied by knife coating, trowel, roller coating, pump and high pressure injection methods.

  • Excellent retention of strength after aging in typical environments
  • Easy application by knife coating, trowel, roller coating, pump and high pressure injection methods
  • Wide process window for curing: 300 to 450 °F (150 to 232 °C)
  • Shear Strength 3200 lbs./sq. in. at Room Temperature
  • Viscosity 40000 - 80000 cP

Widely used for metal to metal bonding, building and home construction, aircraft and automobile manufacturing, bicycle, boat, golf clubs, ski and snowboard assembly.