Orders LinkContactUsLinkComponent
PBS Buffer, Cytiva
Marketsource
Supplier:   Cytiva
PBS Buffer, Cytiva
Supplier:  Cytiva
Marketsource
Certificates
PBS Buffer, Cytiva
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Phosphate buffered saline solution containing detergent, which is recommended as running buffer for small molecule analysis in Biacore systems. Concentrated stock solution, yielding pH 7,4 when diluted 10X.
  • Can be stored at room temperature, precipitation can occur at low temperature
  • Supplemented with DMSO
Specifications for Product Family
Specifications
Specifications
  • Catalog No:
  • 101724-758
  • 103370-302
  • For Use With:
  • Interaction analyses in Biacore systems, most commonly small molecule assays
  • Interaction analyses in Biacore systems, most commonly small molecule assays
Specifications
Product Family Options
Product Information
  • Size
    Availability
    Price
  • 101724-758
    PBS buffer 10X, includes 0,1 M phosphate buffer with 27 mM KCl and 1,37 M NaCl
    PBS buffer 10X, includes 0,1 M phosphate buffer with 27 mM KCl and 1,37 M NaCl
    1000 ml
    Log in for availability
    Log in for availability
    Each
    Each - $337.68
    $337.68
    Each
       $337.68
    Supplier Number: BR100672
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    101724-758
    For Use With
    Interaction analyses in Biacore systems, most commonly small molecule assays
    Marketsource
  • 103370-302
    PBS-P+ buffer 10X, includes 0,2 M phosphate buffer with 27 mM KCl, 1,37 M NaCl and 0,5% surfactant P20 (tween 20).
    PBS-P+ buffer 10X, includes 0,2 M phosphate buffer with 27 mM KCl, 1,37 M NaCl and 0,5% surfactant P20 (tween 20).
    1000 ml
    Log in for availability
    Log in for availability
    Each
    Each - $342.48
    $342.48
    Each
       $342.48
    Supplier Number: 28-9950-84
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    103370-302
    For Use With
    Interaction analyses in Biacore systems, most commonly small molecule assays
    Marketsource
This product has been blocked by your organization. Please contact your purchasing department for more information