Human Type AB Serum, MP Biomedical
Catalog # IC092930149
Supplier: MP Biomedicals
Specifications
- Description:Human Serum, Pooled
- Size:100 ml
- Cat. No.:IC092930149
Specifications
About this item
Human Type AB Serum is used as a growth supplement for cell culture media and in other research applications.
- Collected from Type AB human blood
- Liquid media
- Storage at —20 °C
It has been proven to grow many human cell lines at a faster rate and with a smaller percentage of serum than mixed blood group serum.