Order Entry
Puerto Rico
ContactUsLinkComponent
Trace Element Solution 1, MP Biomedicals
Trace Element Solution 1, MP Biomedicals
Catalog # IC091676549
Supplier:  MP Biomedicals
CAS Number:  
undefined
Trace Element Solution 1, MP Biomedicals
Catalog # IC091676549
Supplier:  MP Biomedicals
Supplier Number:  091676549
CAS Number:  

Specifications

  • Media Format:
    Liquid
  • Description:
    Trace Element Solution 1
  • Size:
    100 ml
  • Storage Temperature:
    4 °C
  • Cat. No.:
    IC091676549

Specifications

About this item

MP Biomedicals offers a 1000X sterile filtered solution for use with serum-free media.

  • Includes selenium, copper, zinc, and iron, as well as ferric citrate for greater stability