Order Entry
United States
ContactUsLinkComponent
RPMI 1640 Medium (1X), MP Biomedical, LLC.
RPMI 1640 Medium (1X), MP Biomedical, LLC.
Catalog # IC091629754
Supplier:  MP Biomedicals
CAS Number:  
RPMI 1640 Medium (1X), MP Biomedical, LLC.
Catalog # IC091629754
Supplier:  MP Biomedicals
Supplier Number:  091629754
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Media Format:
    Liquid
  • Description:
    RPMI 1640 (1X), without L-Glutamine and Phosphate, with 0.85 g/l Sodium Bicarbonate
  • Size:
    500 ml
  • Environmentally Preferable:
  • Storage Temperature:
    4 °C
  • Cat. No.:
    IC091629754

Specifications

About this item

RPMI is now a popular general purpose medium when properly supplemented. As aused modification of McCoy′s 5A Medium, it was formulated to support lymphoblastoid cells in suspension culture, but it has since been shown to support a wide variety of cells that are anchorage dependent.

  • Originally developed by Moore and his colleagues at Roswell Park Memorial Institute (RPMI)
  • Used for the cultivation of normal human and neoplastic leukocytes
  • Widely used in fusion protocols and in the growth of hybrid cells

The Dutch Modification f RPMI 1640 is also available through MP contains 6400 mg/l sodium chloride instead of 6000 mg/l, 1000 mg/l sodium bicarbonate instead of 2000 mg/l, and 20 mM HEPES.

It was based on the RPMI 1630 line of media which utilized a bicarbonate buffering system and alterations in the amounts of amino acids and vitamins.

Originally intended to be used with a serum supplement, RPMI 1640 has been shown to support several cell lines in the absence of serum.