Bio-Rich 1 (DMEM/F12 Enriched), MP Biomedical, LLC.
Catalog # IC091047122
Supplier: MP Biomedicals
CAS Number:
Specifications
- Media Format:Powder
- Description:BioRich 1 (DMEM/F-12 Enriched)
- Size:1× 10 ml
- Storage Temperature:4 °C
- Cat. No.:IC091047122
Specifications
About this item
BioRich 1 is a DMEM/F-12 powdered media blend enriched with trace elements. This permits the transition to 'serum-free' conditions reducing or eliminating the necessity for serum supplementation.
- Used with ITS Premix, a completely serum independent
- Elevated growth of hybridoma lines at reduced serum concentrations
BioRich 1 with 10 common anchorage dependent cell lines commonly show increased yields from 8 to 114% when compared to standard media in normal serum-containing systems. BioRich 1 will support comparable growth to standard media (containing 10% serum) at reduced serum concentrations and in some lines will maintain growth at serum concentrations which will not support growth when using standard media.