Order Entry
Puerto Rico
ContactUsLinkComponent
Tryptic Soy Agar, MP Biomedicals
Tryptic Soy Agar, MP Biomedicals
Catalog # IC091010617
Supplier:  MP Biomedicals
CAS Number:  
undefined
Tryptic Soy Agar, MP Biomedicals
Catalog # IC091010617
Supplier:  MP Biomedicals
Supplier Number:  091010617
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Media Format:
    Powder
  • Description:
    Tryptic Soy Agar
  • Size:
    500 g
  • Cat. No.:
    IC091010617
  • Storage temperature:
    Room Temperature (15...30 °C)

Specifications

About this item

Tryptic Soy Agar is a medium very rich in nutrients for 'general use' in microbiological laboratories. If desired, antibiotics can easily be incorporated as well as other supplements or inhibitory agents.

  • Used for the isolation and cultivation of fastidious, non-fastidious micro-organisms and also for sterility testing
  • Lacks carbohydrates
  • Useful in the study of hemolytic reactions and also in the preparation of chocolate agar
  • Used with or without blood supplement
  • Used as precultivation medium and enumeration (E. coli) according to membrane-filter technique

In Tryptic Soy Agar, casein peptone and soya peptone provide nitrogen, vitamins and minerals. The natural sugars from soya peptone promote bacterial growth. Sodium chloride is for the osmotic balance. As it does not contain the X and V factors, it is suitable for identification of Haemophilus sp. by adding X (Hemin) and V (DPN) factors strips

Suspend 40 g of the medium in one liter of deionized or distilled water. Heat with frequent agitation and boil for one minute. Sterilize in an autoclave between 118 and 121 °C at a pressure of not more than 15 lbs. for 15 minutes. In the case of large volume preparation, increase the time of sterilization but not the temperature or pressure. Cool and pour into petri dishes. For hemolytic studies, add 5 to 10% sheep or rabbit blood.

A short list of microorganisms that grow on this medium are the following: Streptococcus, Neisseria, Brucella, Corynebacterium, Listeria, Pasteurella, Vibrio, Haemophilus vaginals, Candida and more.