HiScreen™ MabSelect™ PrismA Protein A Column, Cytiva
76237-722
: Cytiva
- Particle Size:60 µm
- Max. Pressure:3 bar
- Capacity:4.7 ml
- Description:HiScreen™ MabSelect™ PrismA
- Environmentally Preferable:
- pH Range:3 – 12
- Temperature Limit:40 °C
- Binding Capacity:~ 80 mg human IgG/ml resin at 6 min residence time
~ 40 mg human IgG/ml resin at 2 min residence time - Storage:20% ethanol, 2 to 8 °C
- Column hardware:PP
- Bed dimensions:7.7×100 mm
- Flow velocity:<500 cm/h
- Cat. No.:76237-722
These columns are prepacked with MabSelect™ PrismA protein A chromatographyn resin that has been improved with an optimised high-flow agarose base matrix and a genetically engineered protein A-derived ligand, allowing efficient cleaning between purification runs. These columns are suitable for purifying antibodies with affinity chromatography in research and process development.
- Enhanced binding capacity compared with other Protein A resins
- Excellent alkaline stability enables efficient cleaning and sanitisation using 0.5 to 1.0 M NaOH
- Convenient HiScreen™ format for easy connection to a syringe, peristaltic pump, or chromatography system such as an ÄKTA system
- Convenient method optimisation and parameter screening enabled by the 10 cm bed height
MabSelect™ PrismA builds on the proven track record of MabSelect™ and MabSelect™ SuRe™ resins in commercial mAb production.
The columns are made of biocompatible PP that does not interact with biomolecules.
: Note that HiScreen™ columns cannot be opened or repacked.