Order Entry
Puerto Rico
ContactUsLinkComponent
Egg PC (60%) 25 mg/mL in chloroform
Egg PC (60%) 25 mg/mL in chloroform
Catalog # 100127-830
CAS Number:  
Egg PC (60%) 25 mg/mL in chloroform
Catalog # 100127-830
Supplier Number:  241601C1G
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle, amber, narrow mouth, screw cap, 30 ml
  • Size:
    1 g (2×20 ml)
  • Cat. No.:
    100127-830
  • Supplier no.:
    241601C1G

Specifications

About this item

 :
Storage Temperature:Freezer
CAS Number:97281-44-2