Yeast Extract (Paste), HiMedia Laboratories
Supplier: HiMedia
Certificates
About this item
Yeast extract paste suitable for cultivation of various microorganisms, fermentation and other biological products.
- Brownish yellow to brown homogenous highly viscous paste, having characteristic odor but not putrescent
- Freely soluble in distilled/purified water, insoluble in alcohol
Specifications
- Storage Temperature:<30 °C
- pH:4.70 - 6.70