Order Entry
Puerto Rico
ContactUsLinkComponent
Hanks' Balanced Salts, without Sodium Bicarbonate, MP Biomedical, LLC
Hanks' Balanced Salts, without Sodium Bicarbonate, MP Biomedical, LLC
Supplier:  MP Biomedicals
Certificates
Hanks' Balanced Salts, without Sodium Bicarbonate, MP Biomedical, LLC
Supplier:  MP Biomedicals

Specifications

  • Format:
    Orange Powder
  • pH Range:
    6.0 - 7.0
  • Storage Temperature:
    Room Temperature (15…30 °C).

Specifications

Frequently Bought Together