Order Entry
Puerto Rico
ContactUsLinkComponent
 
StabilBlot™ Casein Blocker-5X, TBS, BioFX®, Surmodics IVD, Inc.
Catalog # 102466-292
Supplier:  Surmodics
CAS Number:  
StabilBlot™ Casein Blocker-5X, TBS, BioFX®, Surmodics IVD, Inc.
Catalog # 102466-292
Supplier:  Surmodics
Supplier Number:  SB05-0125-01
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    StabilBlot™ Casein Blocker-5X, TBS
  • Size:
    125 mL
  • Protein:
    Casein
  • Shelf Life:
    3 years
  • Storage Buffer:
    TBS
  • Storage Temperature:
    2…8 °C
  • Cat. No.:
    102466-292
  • Supplier no.:
    SB05-0125-01

Specifications

About this item

The StabilBlot™ Blocker Family offers a range of options, both protein-based and synthetic-based, to block non-specific binding as well as enhance sensitivity when used as a blocker and antibody diluent.

  • Enhances Sensitivity
  • Minimizes background noise
  • 3 year shelf life

StabilBlot™ Casein Blocker-5X, TBS reduces non-specific binding to enhance sensitivity in Western Blot applications.

Certifications: ISO 13485: 2016 & 9001:2015 Certified.