Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Description:Alkaline Phosphatase/Horseradish Peroxidase Block
- Shelf Life:3 years
- Storage Temperature:2 to 8 °C
- Application:Immunohistochemistry (IHC)
- Cat. No.:100359-236
- Supplier no.:APHP-0100-01
Specifications
About this item
BioFX® Alkaline Phosphatase/Horseradish Peroxidase Block is supplied as a ready-to-use blocking reagent for immunohistochemistry samples. The solution blocks both alkaline phosphatase and peroxidase endogenous enzymatic activity.
- HRP and AP Blocker
- 3 year shelf life
- Ready to use formulation
Alkaline Phosphatase/Horseradish Peroxidase Block blocks the endogenous alkaline phosphatase and horseradish peroxidase present in many immunohistochemistry samples.