Specifications
- Description:DNA Extraction Kit in Wells (96 Deep-Well Plate)
- Size:1 Kit
- Cat. No.:102650-476
- Supplier no.:D100W
Specifications
About this item
The DNA extraction is performed in a 96 deep-well plate, a convenient high-throughput format that allows you to process up to 96 samples or multiple plates in the same run.
Use multichannel pipettors to increase efficiency and decrease repetitive handling while minimizing mistakes associated with handling individual tubes. The proprietary co-precipitating reagent used in the kit keeps the pellet seated in the well and does not interfere with qPCR. Since the kit does not use iodine or glycogen clothes will be stain-free. Each kit has reagent sufficient for 96 individual extractions in less than 2 hours.