Burrs for Micro Drill, Fine Science Tools
Supplier: FINE SCIENCE TOOLS INC MS
Certificates
About this item
Made from stainless steel or carbon steel
- Autoclavable
Shaft diameter: 2.3 mm
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.