BioClean™ Bouffant Caps, BBC, Ansell
75972-518
- Clothing Size:One size
- Color:Blue
- Fabric:PP
- Weight:10 g/m²
- Cat. No.:75972-518
- Supplier no.:BBC-W
The BioClean™ bouffant cap is lightweight and breathable and reduces the risk of contamination from the head area from entering the controlled environment.
- Used in life sciences industry
- Latex-free elastic
- Stitched seams
- Weight: 10 GSM
Soft bouffant caps are made from spunbond polypropylene with elasticated double stitching.
: Caps are packaged 100 per sealed inner PE bag; 10 inner bags per sealed outer PE bag; one outer bag per lined carton (1000 pieces).