ContactUsLinkComponent
VECTABOND® Reagent for Tissue Section Adhesion, Vector Laboratories
VECTABOND® Reagent for Tissue Section Adhesion, Vector Laboratories
Catalog # 101098-506
undefined
VECTABOND® Reagent for Tissue Section Adhesion, Vector Laboratories
Catalog # 101098-506
Supplier Number:  SP-1800-7
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Volume:
    7 ml
  • Description:
    VECTABOND® Reagent for Tissue Section Adhesion
  • Cat. No.:
    101098-506
  • Supplier no.:
    SP-1800
Specifications
About this item
Designed to significantly increase the adherence of both frozen and paraffin-embedded tissue sections and cell preparations to glass slides and coverslips.
  • Chemically modifies the surface of the glass with positive charges to form a highly adherent surface
  • Tissues remain attached even under extreme conditions such as high temperature antigen unmasking techniques and in situ hybridization
  • VECTABOND Reagent treated slides can be stored for long periods
  • 7 ml concentrate dilutes to 350 ml
  • Treats at least 500 standard slides
Applications: Immunohistochemistry / Immunocytochemistry, Immunofluorescence, In situ hybridization.