Organic Reference Standard, Diquat solution, Ultra Scientific
Catalog # ULPST-1410AS
Supplier: Ultra Scientific
Small Business Enterprise
CAS Number:
Specifications
- Concentration:100 µg/ml in water (Low TOC <50 ppb)
- Description:Diquat Standard (from dibromide)
- Size:1×1 ml
- Description:Diquat Standard
- Cat. No.:ULPST-1410AS
- Supplier no.:PST-1410AS