Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Artificial Eccrine Perspiration, Pickeringlab

Artificial Eccrine Perspiration, Pickeringlab

Artificial eccrine perspiration, Bottle, 5 ml
Catalog # 102465-390
Marketsource
Supplier:   Pickering Labs
Supplier #:  1700-0024
Artificial Eccrine Perspiration, Pickeringlab
Catalog # 102465-390
Supplier:  Pickering Labs
Marketsource
Artificial Eccrine Perspiration, Pickeringlab

Artificial eccrine perspiration, Bottle, 5 ml

Price
$133.38
Log in for availability
Each (5ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Artificial eccrine perspiration is a ready-to-use solution and is the closest mimic to true human eccrine sweat.

  • Universally standardize
  • Used in research and product testing
  • Formula that can satisfy all test challenges
  • Reliable, reproducible and consistent solutions

It consists of 19 amino acids, the seven most abundant minerals, and the four most abundant metabolites at a pH of 4.5. All concentrations closely match experimentally determined values for adult human eccrine sweat.

The stabilized solution is preserved with a fungicide and bactericide for two years of shelf life at room temperature.

Specifications
  • Description:
    Artificial eccrine perspiration
  • Size:
    5 ml
  • Packaging:
    Bottle

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.