

Hydrochloric acid 0.01 N in isopropanol (IPA)
Catalog # 103193-766
Supplier: Aqua Solutions
CAS Number:
Specifications
- Pack type:Plastic bottle, clear
- Size:1 L
- Cat. No.:103193-766
- Supplier no.:SPE194
Specifications
About this item
:
ADR:3,II
:36.46
UN:1993
:GHS02
:GHS07
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324