Order Entry
Puerto Rico
ContactUsLinkComponent
Dispase (from Bacillus polymyxa), STEMCELL Technologies
Dispase (from Bacillus polymyxa), STEMCELL Technologies
Catalog # MSPP-7913
CAS Number:  
Dispase (from Bacillus polymyxa), STEMCELL Technologies
Catalog # MSPP-7913
Supplier Number:  07913
CAS Number:  

Specifications

  • Source:
    Bacillus polymyxa
  • Species:
    Human
    Mouse
    Non-human primate
    Rat
  • Size:
    5 units/ml
  • Enzyme Name:
    Dispase (5 U/ml)
  • Enzyme Synonyms:
    Neutral protease; Proteinase
  • Cat. No.:
    MSPP-7913
  • Supplier no.:
    7913

Specifications

About this item

Dispase is a protease that is suitable for the gentle dissociation of a wide variety of tissues.

  • Achieve gentle dissociation in a wide variety of tissues
  • Optimized for for the generation of single-cell suspensions from dissociated human and mouse mammary tissue

It contains 5 U/ml dispase II (neutral protease from Bacillus polymyxa) dissolved in Hanks’ balanced salt solution.

Use Dispase for gentle dissociation of a wide variety of tissues. Incubation of minced tissue with pre-warmed Dispase and gentle agitation will liberate cells with minimal cell damage. Pre-warmed Dispase can also be used to harvest cells from tissue culture plastic. This proteolytic dissociation reagent has been optimized for use in the generation of a single-cell suspension from dissociated human and mouse mammary tissue.

Unlike trypsin, dispase is not inhibited by serum. Dispase activity is inhibited by EDTA and EGTA. Dispase should be removed from cell suspensions by centrifugation followed by washing the cells with buffer or culture medium.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').