Elbow Rest, Nuaire
Catalog # MSPP-NU978002
Supplier: NUAIRE INC MS
Specifications
- Description:Elbow rest
- Size:10"
- Cat. No.:MSPP-NU978002
- Supplier no.:NU-978-002
Specifications
About this item
The rest promote a healthy working environment by improving forearm and elbow support while keeping arms off the front air grill of Class II biological safety cabinet.
- Comfortable close-cell sponge padding does not absorb liquids
- Conveniently placed on front air grill of a biological safety cabinet
- Fits all makes and models of biological safety cabinets
- Easy to clean and autoclavable
- Rubber feet caps included for use on flat surfaces such as countertops
Ergonomic design of the elbow rest keeps your elevated above the front grill so airflows are not disrupted yet keeps your arms and shoulder in a comfortable position to perform your work.