- Description:ExoQuick Plasma Precipitation Kit with Thrombin
- Size:75 reactions
- Environmentally Preferable:
- Cat. No.:MSPP-EXOQ5TM1
- Supplier no.:EXOQ5TM-1
A single kit optimizes exosome isolation from plasma by combining thrombin for de-fibrination with ExoQuick.
- Saves time and labor
- Easily scalable
- Conserves precious sample
- Delivers high yields of functional, high quality exosomes
Can be used to isolate exosomes for a wide range of downstream applications including biomarker studies exosomal miRNA profiling, exosomal proteomics, exosomal lipidomics/metabolomics, functional studies such as in cell-to-cell signaling, basic biology such as role in tumorigenesis.