Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Trypsin

Trypsin

100 ml
Catalog # MSPP-30-2101
Marketsource
Supplier #:  30-2101
Trypsin
Catalog # MSPP-30-2101
Marketsource
CAS Number:  
Trypsin

100 ml

CAS Number:  
Price
$27.69
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

This solution contains 0.25% Trypsin/0.53 mM EDTA in Hanks balanced salt solution without calcium or magnesium. For dissociation of cell monolayers.

Trypsin-EDTA solution is suitable for most but not for all adherent cell lines. This product contains phenol red.

Specifications

Specifications

  • Size:
    100 ml
  • Storage Conditions:
    Frozen
  • Cat. No.:
    MSPP-30-2101
  • Shipping Conditions:
    Frozen
  • Enzyme Name:
    Trypsin

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.