ContactUsLinkComponent
Torch Bonnet, PerkinElmer
Torch Bonnet, PerkinElmer
Catalog # 97012-906
Supplier:  PerkinElmer
undefined
Torch Bonnet, PerkinElmer
Catalog # 97012-906
Supplier:  PerkinElmer
Supplier Number:  N0775289
Specifications
  • Description:
    Quartz Bonnet
  • Cat. No.:
    97012-906
  • Supplier no.:
    N0775289
Specifications
About this item
This quartz bonnet is used with Optima™ 2x00 DV, 4x00 DV, 5x00 DV, 7x00 DV ICP-OES series instruments.