Order Entry
Puerto Rico
ContactUsLinkComponent
Clamp, Three Pronged, Swivel, Chemglass
Clamp, Three Pronged, Swivel, Chemglass
Catalog # 80063-612
Supplier:  Chemglass
Small Business Enterprise
undefined
Clamp, Three Pronged, Swivel, Chemglass
Catalog # 80063-612
Supplier:  Chemglass
Supplier Number:  CG-9215-01
Small Business Enterprise

Specifications

  • Description:
    Clamps, three prong, swivel
  • Extension arm length:
    1.6 cm
  • Cat. No.:
    80063-612
  • Max. grip size:
    69 mm (2.72")
  • Grip size:
    1.9 cm
  • Supplier no.:
    CG-9215-01

Specifications

About this item

Dual adjustment clamp with shaft wing nut to allow clamp jaw movement of 360°.

Delivery: Supplied with removable vinyl and fiberglass covers.