Universal Vials, Ab Sciex
Catalog # MSPP-A62251
Supplier: AB SCIEX LLC MS
Specifications
- Cap included:No
- Description:Universal Vials
- Material:Polymethylpentene
- Environmentally Preferable:
- Cat. No.:MSPP-A62251
- Supplier no.:A62251
Specifications
About this item
Universal vials are used to accommodate buffers/samples.
These polymethylpentene vials are precision manufactured and have been compatibility tested with commonly used CE reagents. Universal vials are designed for use only with CESI 8000, PA 800 Enhanced or PA 800 plus instruments and should not be reused.