ContactUsLinkComponent
Nitrate Reducing Agent
  470225-858
 :  LaMotte
 :  
undefined
Nitrate Reducing Agent
  470225-858
 :  LaMotte
 :  V-6279-C
 :  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    Nitrate reducing agent
  • Size:
    5 g
  • Cat. No.:
    470225-858
  • Supplier no.:
    V-6279-C
Nitrate reducing agent for LaMotte products.
  • Precise nitrate testing: Provides accurate nitrate reduction measurements, essential for water quality analysis
  • Consistent results: Delivers reliable and repeatable results for educational and scientific purposes
  • Compatible with LaMotte kits: Designed to integrate seamlessly with LaMotte’s water testing kits for easy use
The nitrate reducing reagent from LaMotte is a perfect classroom tool for hands-on environmental science experiments. Ideal for exploring water quality, it allows students to investigate how nitrates impact ecosystems and water treatment. Easy to use and reliable, it brings real-world science into your lessons, helping students connect theory with practical applications.