Chlorine Colorimeter, DC155-CL, LaMotte®
Catalog # 470308-446
Supplier: LaMotte
CAS Number:
Specifications
- Chemical method:DPD (N,N-diethyl-p-phenylenediamine)
- Description:Chlorine Colorimeter
- Size:1
- Model:DC-1500-CL
- Sample Cell Compatability:25 mm diameter flat bottom test tubes
- User Programs:Pre-programmed test, absorbance, %T
- Accuracy:±2% FS
- Automatic Switch-Off:Disabled, 5, 10, 50 minutes
- Compliance:IP67
- Display:160 x 100 backlit LCD, 20 x 6 line graphical
- Keypad:6 button
- Optics:LED, silicon photodiode with integrated interference
- Power supply:Lithium ion rechargeable battery, 3.7v, 2.5 x .75
- Cat. No.:470308-446
- Range:0 - 4 ppm
Specifications
About this item
The Lamotte Chlorine Colorimeter Kit, ideal for both field and lab use, covers the entire critical chlorine range of 0-4 ppm with an MDL of 0.03 ppm.
- Chlorine Colorimeter Kit - DPD Tablet
- Large graphical liquid crystal display
- Simple, menu-driven operation
- Lithium ion rechargeable battery and computer/wall adapter
- IP67 Waterproof